missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse TRAIL/TNFSF10 Recombinant Protein

$404.12 - $404.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22035
Name TRAIL/TNFSF10
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
1121TL010 R&D Systems
RDS1121TL010
Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4 with BSA as a carrier protein. 20 kda Each for $404.12
1121TL010CF R&D Systems
RDS1121TL010CF
Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4. 20 kDa (monomer) Each for $404.12
Specifications

Description

Source: E. coli-derived, Pro118-Asn291
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Tris, (NH4)2SO4 and ZnSO4.
Entrez Gene ID: 22035
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
TRAIL/TNFSF10
10μg
Mouse
22035
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes