Please login to your online account to display your discounted pricing

R&D Systems Mouse TNF RI/TNFRSF1A Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 21937
Molecular Weight 21 kDa
Name TNF RI/TNFRSF1A
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

425R1050

 
r&d systems
RDS425R1050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94

425R1050CF

 
r&d systems
RDS425R1050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 21937
Molecular Weight 21 kDa
Name TNF RI/TNFRSF1A
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Ile22-Ala212, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21937
Regulatory Status: RUO