missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse TNF-alpha aa 80-235 Recombinant Protein, N-terminal Met

$259.40 - $819.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 21926
Molecular Weight 17 kDa
Name TNF-alpha aa 80-235
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
410MT050 R&D Systems
RDS410MT050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $819.94
410MT010CF R&D Systems
RDS410MT010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $259.40
410MT010 R&D Systems
RDS410MT010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $259.40
410MT050CF R&D Systems
RDS410MT050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $819.94
Specifications

Description

Source: E. coli-derived, Leu80-Leu235, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21926
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
21926
TNF-alpha aa 80-235
Yes