missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Sonic Hedgehog/Shh N-Terminus Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20423
Molecular Weight 20 kDa
Name Sonic Hedgehog/Shh N-Terminus
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
461SH025 R&D Systems
RDS461SH025
Lyophilized from a 0.2μm filtered solution in PBS, Trehalose and with BSA as a carrier protein. Each for $376.73
461SH025CF R&D Systems
RDS461SH025CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $376.73
Specifications

Description

Source: E. coli-derived, Cys25-Gly198, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 20423
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20423
Sonic Hedgehog/Shh N-Terminus
25μg
Mouse