missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Sonic Hedgehog/Shh (C25II), N-Terminus Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20423
Molecular Weight 19.8 kDa
Name Sonic Hedgehog/Shh (C25II), N-Terminus
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
464SH025 R&D Systems
RDS464SH025
Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and DTT with BSA as a carrier protein. 25μg Each for $415.83
464SH025CF R&D Systems
RDS464SH025CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and DTT . 25μg Each for $415.83
Specifications

Description

Source: E. coli-derived, Cys25-Gly198 (Cys25Ile-Ile), with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and DTT .
Entrez Gene ID: 20423
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20423
Sonic Hedgehog/Shh (C25II), N-Terminus
Yes