missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Soggy-1/DkkL1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 50722
Molecular Weight 26.8 kDa
Name Soggy-1/DkkL1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1508S1025 R&D Systems
RDS1508S1025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1508S1025CF R&D Systems
RDS1508S1025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu21-Leu230, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 50722
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
26.8 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
50722
Soggy-1/DkkL1
25μg
Mouse