missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse sFRP-3 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20378
Molecular Weight 34 kDa
Name sFRP-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
592FR010 R&D Systems
RDS592FR010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
592FR010CF R&D Systems
RDS592FR010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala33-Ser323 with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 34 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20378
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
34 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20378
sFRP-3
10μg
Mouse