Please login to your online account to display your discounted pricing

R&D Systems Mouse Semaphorin 3A Fc Chimera Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 110.1 kDa (monomer)
Name Semaphorin 3A Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

5926S3025

 
r&d systems
RDS5926S3025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

5926S3025CF

 
r&d systems
RDS5926S3025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 110.1 kDa (monomer)
Name Semaphorin 3A Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: Chinese Hamster Ovary cell line, CHO-derived, Asn21-Lys747: Arg552Ala, Arg554Ala, Arg555Ala, Arg618Ala, Lys619Ala
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 110.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO