missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Ret Fc Chimera Recombinant Protein

$454.94 - $454.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 19713
Molecular Weight 95 kDa (monomer)
Name Ret Fc Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
482RT100 R&D Systems
RDS482RT100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $454.94
482RT100CF R&D Systems
RDS482RT100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $454.94
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu29-Arg637 (Phe174Ser)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 95 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 19713
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
95 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
19713
Ret Fc Chimera
100μg
Mouse