missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Relaxin-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 19773
Name Relaxin-1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
6637RN025 R&D Systems
RDS6637RN025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 3.9 kDa (B chain) & 2.7 kDa (A chain) Each for $428.87
6637RN025CF R&D Systems
RDS6637RN025CF
Lyophilized from a 0.2μm filtered solution in PBS. 3.7 kDa (B chain) & 2.7 kDa (A chain) Each for $428.87
Specifications

Description

Source: E. coli-derived, Arg23-Arg54, with an N-terminal Met (B chain) & Glu161-Cys185 (A chain)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 3.7 kDa (B chain) & 2.7 kDa (A chain)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 19773
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
Relaxin-1
25μg
Mouse
19773
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes