missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Reelin Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 19699
Molecular Weight 162.1 kDa
Name Reelin
Purity 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3820MR025 R&D Systems
RDS3820MR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
3820MR025CF R&D Systems
RDS3820MR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu1221-Ile2661, with a C-terminal 6-His tag
Purity: 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 162.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 19699
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
162.1 kDa
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
19699
Reelin
25μg
Mouse