missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse R-Spondin 3 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 72780
Molecular Weight 28.9 kDa
Name R-Spondin 3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4120RS025CF R&D Systems
RDS4120RS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
4120RS025 R&D Systems
RDS4120RS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-His277
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 28.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 72780
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
28.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
72780
R-Spondin 3
25μg
Mouse