missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse R-Spondin 1 Recombinant Protein, (CHO-expressed)

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 192199
Molecular Weight 27 kDa
Name R-Spondin 1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7150RS025 R&D Systems
RDS7150RS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
7150RS025CF R&D Systems
RDS7150RS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-Gln265
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 27 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 192199
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
27 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
192199
R-Spondin 1
25μg
Mouse