missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse PlGF-2 Recombinant Protein

$366.12 - $1,113.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 18654
Molecular Weight 15.1 kDa (monomer)
Name PlGF-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
465PL010 R&D Systems
RDS465PL010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $366.12
465PL010CF R&D Systems
RDS465PL010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $366.12
465PL050 R&D Systems
RDS465PL050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $1,113.55
465PL050CF R&D Systems
RDS465PL050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $1,113.55
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala24-Pro158 & Ala27-Pro158
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 18654
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.1 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
18654
PlGF-2
Yes