missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse P-Selectin/CD62P Fc Chimera Recombinant Protein

$298.51 - $885.12

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 20344
Molecular Weight 99.1 kDa (monomer)
Name P-Selectin/CD62P Fc Chimera
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
737PS050 R&D Systems
RDS737PS050
50μg Each for $298.51
737PS200 R&D Systems
RDS737PS200
200μg Each for $885.12
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Trp42-Ala709
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 99.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20344
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20344
P-Selectin/CD62P Fc Chimera
Yes
Lyophilized from a 0.2μm filtered solution in PBS.
99.1 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse