missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse OX40 Ligand/TNFSF4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22164
Molecular Weight 18 kDa
Name OX40 Ligand/TNFSF4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1236OX025 R&D Systems
RDS1236OX025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1236OX025CF R&D Systems
RDS1236OX025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln49-Leu198, with an N-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22164
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
22164
OX40 Ligand/TNFSF4
25μg
Mouse