missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Osteopontin/OPN Recombinant Protein

$302.77 - $885.52

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20750
Molecular Weight 31.5 kDa
Name Osteopontin/OPN
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
441OP050 R&D Systems
RDS441OP050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $302.77
441OP050CF R&D Systems
RDS441OP050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $302.77
441OP200 R&D Systems
RDS441OP200
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 200μg Each for $885.52
441OP200CF R&D Systems
RDS441OP200CF
Lyophilized from a 0.2μm filtered solution in PBS. 200μg Each for $885.52
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu17-Asn294 (Glu99Gly), with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 31.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20750
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
31.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20750
Osteopontin/OPN
Yes