Please login to your online account to display your discounted pricing

R&D Systems Mouse Netrin-G1a Recombinant Protein


Manufacturer: r&d systems  RDS1166NG025CF

 View more versions of this product

Catalog No. 1166NG025CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 56 kDa
Name Netrin-G1a
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 80883