Please login to your online account to display your discounted pricing

R&D Systems Mouse Netrin-4 Recombinant Protein


Manufacturer: r&d systems  RDS1132N4025CF

 View more versions of this product

Catalog No. 1132N4025CF

  • / Each
 

Description & Specifications

Specifications

Recombinant Yes
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Name Netrin-4
Gene ID (Entrez) 57764
Molecular Weight 69 kDa
Source Mouse
Purity 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Conjugate Unlabeled