missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Netrin-4 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 57764
Molecular Weight 69 kDa
Name Netrin-4
Purity 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1132N4025 R&D Systems
RDS1132N4025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1132N4025CF R&D Systems
RDS1132N4025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu19-Val628 & Ala26-Val628, both with a C-terminal 10-His tag
Purity: 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 69 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 57764
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
69 kDa
80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
57764
Netrin-4
25μg
Mouse