Please login to your online account to display your discounted pricing

R&D Systems Mouse Netrin-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 18208
Molecular Weight 68.2 kDa
Name Netrin-1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1109N1025CF

 
r&d systems
RDS1109N1025CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $428.87

1109N1025

 
r&d systems
RDS1109N1025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 18208
Molecular Weight 68.2 kDa
Name Netrin-1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Val22-Ala603, with a C-terminal 10 His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 68.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Entrez Gene ID: 18208
Regulatory Status: RUO