missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse MSP/MST1 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 15235
Molecular Weight 53.7 kDa ( alpha-chain) & 25 kDa (beta-chain)
Name MSP/MST1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6244MS025CF R&D Systems
RDS6244MS025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
6244MS025 R&D Systems
RDS6244MS025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-Glu716 (Cys677Ala)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 53.7 kDa ( alpha-chain) & 25 kDa (beta-chain)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 15235
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
53.7 kDa ( alpha-chain) & 25 kDa (beta-chain)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
15235
MSP/MST1
25μg
Mouse