missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Meteorin Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 70083
Molecular Weight 30 kDa
Name Meteorin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3475MN025 R&D Systems
RDS3475MN025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
3475MN025CF R&D Systems
RDS3475MN025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gly22-Asp291 with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 30 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 70083
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
30 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
70083
Meteorin
25μg
Mouse