missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse MD-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 17084
Molecular Weight 18.5 kDa
Name MD-1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
130MD050CF R&D Systems
RDS130MD050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
130MD050 R&D Systems
RDS130MD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asp20-Ser16, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 17084
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.5 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
17084
MD-1
50μg
Mouse