Please login to your online account to display your discounted pricing

R&D Systems Mouse MD-1 Recombinant Protein


Manufacturer: r&d systems  RDS130MD050CF

 View more versions of this product

Catalog No. 130MD050CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 18.5 kDa
Name MD-1
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Mouse
Gene ID (Entrez) 17084
Quantity 50μg
Recombinant Yes
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.