missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse MBL-1 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 17194
Molecular Weight 24 kDa
Name MBL-1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2077MB050 R&D Systems
RDS2077MB050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
2077MB050CF R&D Systems
RDS2077MB050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser18-Ala239
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 17194
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
24 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
17194
MBL-1
50μg
Mouse