Please login to your online account to display your discounted pricing

R&D Systems Mouse Matrilin-2 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Molecular Weight 104.9 kDa
Name Matrilin-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

3234MN050

 
r&d systems
RDS3234MN050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

3234MN050CF

 
r&d systems
RDS3234MN050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 104.9 kDa
Name Matrilin-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Arg24-Arg956 & Met40-Arg956, both with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 104.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO