Please login to your online account to display your discounted pricing

R&D Systems Mouse Lymphotoxin α2/β1 Recombinant Protein


Manufacturer: r&d systems  RDS1008LY010

 View more versions of this product

Catalog No. 1008LY010

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
Recombinant Yes
Gene ID (Entrez) 16992
Quantity 10μg
Structural Form Noncovalently-linked heterotrimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Name Lymphotoxin α2/β1
Source Mouse