missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Lymphotoxin α2/β1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16992
Molecular Weight 18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
Name Lymphotoxin α2/β1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1008LY010CF R&D Systems
RDS1008LY010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
1008LY010 R&D Systems
RDS1008LY010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Leu34-Leu202 (LT alpha) & Gly136-Gly306 (LT beta)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16992
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16992
Lymphotoxin α2/β1
10μg
Mouse