missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse LRP-5 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16973
Molecular Weight 152.1 kDa
Name LRP-5
Purity 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7344LR025 R&D Systems
RDS7344LR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
7344LR025CF R&D Systems
RDS7344LR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Met1-Ser1383, with a C-terminal 6-His tag
Purity: 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 152.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16973
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
152.1 kDa
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16973
LRP-5
25μg
Mouse