Please login to your online account to display your discounted pricing

R&D Systems Mouse Limitin/IFN-zeta Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 319146
Molecular Weight 19 kDa
Name Limitin/IFN-zeta
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1535LM010

 
r&d systems
RDS1535LM010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

1535LM010CF

 
r&d systems
RDS1535LM010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 319146
Molecular Weight 19 kDa
Name Limitin/IFN-zeta
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Leu22-Arg182, with a C-terminal Ile and 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 319146
Regulatory Status: RUO