missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Leptin/OB Recombinant Protein

$264.77 - $657.49

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Gene ID (Entrez) 16846
Molecular Weight 16 kDa
Name Leptin/OB
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
498OB01M R&D Systems
RDS498OB01M
1mg Each for $264.77
498OB05M R&D Systems
RDS498OB05M
5mg Each for $657.49
Specifications

Description

Source: E. coli-derived, Val22-Cys167, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 16846
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16846
Leptin/OB
Yes
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
16 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse