Please login to your online account to display your discounted pricing

R&D Systems Mouse LDL R Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16835
Molecular Weight 85.7 kDa
Name LDL R
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2255LD025

 
r&d systems
RDS2255LD025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

2255LD025CF

 
r&d systems
RDS2255LD025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16835
Molecular Weight 85.7 kDa
Name LDL R
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Ala22-Arg790 (Ala23Val and Cys27Gly), with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 85.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16835
Regulatory Status: RUO