missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse LBP Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16803
Molecular Weight 51.5 kDa
Name LBP
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6635LP025CF R&D Systems
RDS6635LP025CF
Lyophilized from a 0.2μm filtered solution in MES, NaCl, PEG and CHAPS. Each for $428.87
6635LP025 R&D Systems
RDS6635LP025
Lyophilized from a 0.2μm filtered solution in MES, NaCl, PEG and CHAPS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gly25-Val481 (Ser102Arg, Tyr284His), with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 51.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MES, NaCl, PEG and CHAPS.
Entrez Gene ID: 16803
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
51.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
16803
LBP
25μg
Mouse