missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-9 Recombinant Protein

$415.83 - $1,192.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16198
Molecular Weight 14.2 kDa
Name IL-9
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
409ML010CF R&D Systems
RDS409ML010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $415.83
409ML010 R&D Systems
RDS409ML010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $415.83
409ML050CF R&D Systems
RDS409ML050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $1,192.76
409ML050 R&D Systems
RDS409ML050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $1,192.76
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gln19-Pro144
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16198
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
14.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16198
IL-9
Yes