missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-7 Recombinant Protein

$298.51 - $950.30

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16196
Molecular Weight 15 kDa
Name IL-7
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
407ML005CF R&D Systems
RDS407ML005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $298.51
407ML005 R&D Systems
RDS407ML005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $298.51
407ML025CF R&D Systems
RDS407ML025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $950.30
407ML025 R&D Systems
RDS407ML025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $950.30
Specifications

Description

Source: E. coli-derived, Glu26-Ile154, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16196
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16196
IL-7
Yes