missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-6 Recombinant Protein

$311.55 - $997.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16193
Molecular Weight 21.8 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
406ML005CF R&D Systems
RDS406ML005CF
Supplied as a 0.2μm filtered solution in Sodium Acetate and EDTA. 5μg Each for $311.55
406ML025 R&D Systems
RDS406ML025
Lyophilized from a 0.2μm filtered solution in Sodium Acetate and EDTA with Trehalose and with BSA as a carrier protein. 25μg Each for $997.23
406ML005 R&D Systems
RDS406ML005
Lyophilized from a 0.2μm filtered solution in Sodium Acetate and EDTA with Trehalose and with BSA as a carrier protein. 5μg Each for $311.55
406ML025CF R&D Systems
RDS406ML025CF
Supplied as a 0.2μm filtered solution in Sodium Acetate and EDTA. 25μg Each for $997.23
Specifications

Description

Source: E. coli-derived, Phe25-Thr211, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21.8 kDa
Formulation: Supplied as a 0.2μm filtered solution in Sodium Acetate and EDTA.
Entrez Gene ID: 16193
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16193
IL-6
Yes