missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-5 Recombinant Protein

$378.79 - $1,159.15

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16191
Molecular Weight 13.1 kDa (monomer)
Name IL-5
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
405ML005CF R&D Systems
RDS405ML005CF
Supplied as a 0.2μm filtered solution in PBS. 5μg Each for $378.79
405ML005 R&D Systems
RDS405ML005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $378.79
405ML025CF R&D Systems
RDS405ML025CF
Supplied as a 0.2μm filtered solution in PBS. 25μg Each for $1,159.15
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met21-Gly133
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.1 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in PBS.

Entrez Gene ID: 16191
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
13.1 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
16191
IL-5
Yes