missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-36 alpha/IL-1F6 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 54448
Molecular Weight 21 kDa
Name IL-36 alpha/IL-1F6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2297ML025CF R&D Systems
RDS2297ML025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
2297ML025 R&D Systems
RDS2297ML025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: E. coli-derived, Met1-His160
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 54448
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
54448
IL-36 alpha/IL-1F6
25μg
Mouse