missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-28A/IFN-lambda 2 Recombinant Protein

$404.12 - $404.12

Specifications

Conjugate Unlabeled
Molecular Weight 20.5 kDa
Name IL-28A/IFN-lambda 2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4635ML025CF R&D Systems
RDS4635ML025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $404.12
4635ML025 R&D Systems
RDS4635ML025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $404.12
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asp20-Val193, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-28A/IFN-lambda 2
25μg
Mouse
20.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes