missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-27 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 246779
Molecular Weight 48.7 kDa
Name IL-27
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2799ML010 R&D Systems
RDS2799ML010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
2799ML010CF R&D Systems
RDS2799ML010CF
Supplied as a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 48.7 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 246779
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
48.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
246779
IL-27
10μg
Mouse