missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-27 p28/IL-30 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 246779
Molecular Weight 24.9 kDa
Name IL-27 p28/IL-30
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7430ML010CF R&D Systems
RDS7430ML010CF
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS. Each for $416.79
7430ML010 R&D Systems
RDS7430ML010
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Phe29-Ser234, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS.
Entrez Gene ID: 246779
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
24.9 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
246779
IL-27 p28/IL-30
10μg
Mouse