missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-24 Recombinant Protein, (NS0-expressed)

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 93672
Molecular Weight 18 kDa (monomer)
Name IL-24
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
7807ML010 R&D Systems
RDS7807ML010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $376.73
7807ML010CF R&D Systems
RDS7807ML010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $376.73
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln66-Leu220
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 93672
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
93672
IL-24
10μg
Mouse