missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-23 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 83430
Molecular Weight 57 kDa
Name IL-23
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1887ML010 R&D Systems
RDS1887ML010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
1887ML010CF R&D Systems
RDS1887ML010CF
Supplied as a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Met23-Ser335 (p40) & Leu20-Ala196 (p19)
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 57 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 83430
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
57 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
83430
IL-23
10μg
Mouse