Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-22 Recombinant Protein

$407.05 - $407.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 50929
Molecular Weight 16.5 kDa
Name IL-22
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

582ML010

 
r&d systems
RDS582ML010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $407.05

582ML010CF

 
r&d systems
RDS582ML010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 50929
Molecular Weight 16.5 kDa
Name IL-22
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Leu34-Val179, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 50929
Regulatory Status: RUO