Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-20 Recombinant Protein

$432.57 - $432.57

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 58181
Name IL-20
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity    

1204ML025CF

 
r&d systems
RDS1204ML025CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. 17 kDa Each for $432.57

1204ML025

 
r&d systems
RDS1204ML025
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. 17.7 kDa Each for $432.57
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 58181
Name IL-20
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Leu25-Leu176, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 58181
Regulatory Status: RUO