Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-2 (Cys160Ser) Recombinant Protein

$266.69 - $266.69

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16183
Molecular Weight 17 kDa
Name IL-2 (Cys160Ser)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1150ML020

 
r&d systems
RDS1150ML020
Lyophilized from a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. Each for $266.69

1150ML020CF

 
r&d systems
RDS1150ML020CF
Lyophilized from a 0.2μm filtered solution in Sodium Acetate. Each for $266.69
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16183
Molecular Weight 17 kDa
Name IL-2 (Cys160Ser)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Ala21-Gln169 (Cys160Ser), with and without an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 16183
Regulatory Status: RUO