Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-17E/IL-25 Recombinant Protein, (NS0-expressed)

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 140806
Molecular Weight 17.5 kDa (monomer)
Name IL-17E/IL-25
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7909IL010

 
r&d systems
RDS7909IL010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $428.87

7909IL010CF

 
r&d systems
RDS7909IL010CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 140806
Molecular Weight 17.5 kDa (monomer)
Name IL-17E/IL-25
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Val17-Ala169
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 140806
Regulatory Status: RUO