Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-17A/F Heterodimer Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Molecular Weight 14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
Name IL-17A/F Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

5390IL025

 
r&d systems
RDS5390IL025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $441.90

5390IL025CF

 
r&d systems
RDS5390IL025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $441.90
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
Name IL-17A/F Heterodimer
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO