Please login to your online account to display your discounted pricing

R&D Systems Mouse IL-17 RD/SEF Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 171463
Molecular Weight 31.6 kDa
Name IL-17 RD/SEF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2276ML050

 
r&d systems
RDS2276ML050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

2276ML050CF

 
r&d systems
RDS2276ML050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 171463
Molecular Weight 31.6 kDa
Name IL-17 RD/SEF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Gly28-Arg299, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 31.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 171463
Regulatory Status: RUO